Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGTTCCTTTTTCATTAGTGAC[A/C]CCTTCTGAAGTCTTCTTGCCCATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP5 - FK506 binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145775.2 | 1466 | Silent Mutation | GGG,GGT | G,G 436 | NP_001139247.1 | |
NM_001145776.1 | 1466 | Silent Mutation | GGG,GGT | G,G 436 | NP_001139248.1 | |
NM_001145777.1 | 1466 | Intron | NP_001139249.1 | |||
NM_004117.3 | 1466 | Silent Mutation | GGG,GGT | G,G 436 | NP_004108.1 |