Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCATGTCCGAGTCTGAGTCTGA[C/G]ACTGGGTGGAAGCCATAGACGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC26A8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC26A8 - solute carrier family 26 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193476.1 | 2957 | Missense Mutation | TCT,TGT | S,C 941 | NP_001180405.1 | |
NM_052961.3 | 2957 | Missense Mutation | TCT,TGT | S,C 941 | NP_443193.1 | |
NM_138718.2 | 2957 | Missense Mutation | TCT,TGT | S,C 836 | NP_619732.2 | |
XM_011514294.2 | 2957 | Missense Mutation | TCT,TGT | S,C 915 | XP_011512596.1 | |
XM_017010235.1 | 2957 | Missense Mutation | TCT,TGT | S,C 941 | XP_016865724.1 |