Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCACTGTCAGCAGGGCCTGGCTC[A/G]TCCACTCCATTTCTGGAATTATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611687 MIM: 611689 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHDC3L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KHDC3L - KH domain containing 3 like, subcortical maternal complex member | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OOEP - oocyte expressed protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080507.2 | 206 | Missense Mutation | ACG,ATG | T,M 76 | NP_001073976.1 | |
XM_017010890.1 | 206 | Missense Mutation | ACG,ATG | T,M 37 | XP_016866379.1 |
OOEP-AS1 - OOEP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |