Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTGATGTTTCTTAAGAAGTGTC[C/T]GCTTACTAAAGAGTTTACTGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610794 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZSCAN31 - zinc finger and SCAN domain containing 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135215.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | NP_001128687.1 | |
NM_001135216.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | NP_001128688.1 | |
NM_001243241.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | NP_001230170.1 | |
NM_001243242.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 232 | NP_001230171.1 | |
NM_001243243.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 232 | NP_001230172.1 | |
NM_001243244.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 232 | NP_001230173.1 | |
NM_030899.4 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | NP_112161.3 | |
NM_145909.2 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | NP_665916.1 | |
XM_005249295.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_005249352.1 | |
XM_005249296.4 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_005249353.1 | |
XM_011514807.2 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_011513109.1 | |
XM_011514808.2 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_011513110.1 | |
XM_011514809.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_011513111.1 | |
XM_011514811.2 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_011513113.1 | |
XM_011514812.2 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_011513114.1 | |
XM_011514813.2 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_011513115.1 | |
XM_017011196.1 | 1408 | Missense Mutation | CAG,CGG | Q,R 391 | XP_016866685.1 |