Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCAGACTGAAACGCAGTAATCC[A/C]CTGGCAAGGGAAATCTTCCGAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602246 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOB2P1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOB2P1 - transducer of ERBB2, 2 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN9 - zinc finger and SCAN domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199479.1 | 775 | Silent Mutation | CCA,CCC | P,P 45 | NP_001186408.1 | |
NM_001199480.1 | 775 | Silent Mutation | CCA,CCC | P,P 45 | NP_001186409.1 | |
NM_006299.4 | 775 | Silent Mutation | CCA,CCC | P,P 45 | NP_006290.1 | |
XM_011514875.2 | 775 | Silent Mutation | CCA,CCC | P,P 177 | XP_011513177.1 | |
XM_011514876.2 | 775 | Silent Mutation | CCA,CCC | P,P 177 | XP_011513178.1 | |
XM_011514878.2 | 775 | Silent Mutation | CCA,CCC | P,P 177 | XP_011513180.1 | |
XM_017011267.1 | 775 | Silent Mutation | CCA,CCC | P,P 45 | XP_016866756.1 |