Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAATGTCTCCAGGCAAACTGTTC[C/G]TTCACGTAGCCTCGGGACTTGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603632 | ||||||||||||||||||||
Literature Links: |
RPS10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPS10 - ribosomal protein S10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014.4 | 271 | Intron | NP_001005.1 | |||
NM_001203245.2 | 271 | Intron | NP_001190174.1 | |||
NM_001204091.1 | 271 | Intron | NP_001191020.1 |
RPS10-NUDT3 - RPS10-NUDT3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202470.2 | 271 | Missense Mutation | AAC,AAG | N,K 59 | NP_001189399.1 |