Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCCCACACAAAAAGTATGATAA[C/T]CTCTGTCACACATATCACAGAACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616987 MIM: 613069 | ||||||||||||||||||||
Literature Links: |
C6orf120 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf120 - chromosome 6 open reading frame 120 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029863.2 | 3063 | UTR 3 | NP_001025034.1 | |||
NM_001317342.1 | 3063 | UTR 3 | NP_001304271.1 |
PHF10 - PHD finger protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018288.3 | 3063 | Missense Mutation | GAT,GGT | D,G 457 | NP_060758.2 | |
NM_133325.2 | 3063 | Missense Mutation | GAT,GGT | D,G 455 | NP_579866.2 | |
XM_017010998.1 | 3063 | Missense Mutation | GAT,GGT | D,G 405 | XP_016866487.1 |
WDR27 - WD repeat domain 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |