Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCAGCACCTCCACTCTGCACA[C/T]GTAGATGCTGGCGTCATGGCCTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109170 MIM: 600978 MIM: 611550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LST1 - leukocyte specific transcript 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166538.1 | 529 | Intron | NP_001160010.1 | |||
NM_007161.3 | 529 | Intron | NP_009092.3 | |||
NM_205837.2 | 529 | Intron | NP_995309.2 | |||
NM_205838.2 | 529 | Intron | NP_995310.2 | |||
NM_205839.2 | 529 | Intron | NP_995311.2 | |||
NM_205840.2 | 529 | Intron | NP_995312.2 | |||
XM_006715206.3 | 529 | Intron | XP_006715269.1 | |||
XM_006715209.3 | 529 | Intron | XP_006715272.1 | |||
XM_006715210.3 | 529 | Intron | XP_006715273.1 | |||
XM_011514914.2 | 529 | Intron | XP_011513216.1 |
LTB - lymphotoxin beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCR3 - natural cytotoxicity triggering receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145466.1 | 529 | Missense Mutation | ATG,GTG | M,V 107 | NP_001138938.1 | |
NM_001145467.1 | 529 | Missense Mutation | ATG,GTG | M,V 107 | NP_001138939.1 | |
NM_147130.2 | 529 | Missense Mutation | ATG,GTG | M,V 107 | NP_667341.1 | |
XM_006715049.3 | 529 | Missense Mutation | ATG,GTG | M,V 107 | XP_006715112.1 | |
XM_011514459.2 | 529 | Missense Mutation | ATG,GTG | M,V 82 | XP_011512761.1 |