Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCTTACCATTTGCATCTTTTGA[C/A]CAATTGTCTTTGGGATCCATTGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609642 | ||||||||||||||||||||
Literature Links: |
TARP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TARP - TCR gamma alternate reading frame protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003799.1 | 423 | UTR 3 | NP_001003799.1 | |||
NM_001003806.1 | 423 | Missense Mutation | TGG,TGT | W,C 59 | NP_001003806.1 | |
XM_011515407.2 | 423 | Missense Mutation | TGG,TGT | W,C 59 | XP_011513709.1 | |
XM_017012226.1 | 423 | Missense Mutation | TGG,TGT | W,C 59 | XP_016867715.1 | |
XM_017012227.1 | 423 | Missense Mutation | TGG,TGT | W,C 59 | XP_016867716.1 | |
XM_017012228.1 | 423 | UTR 5 | XP_016867717.1 | |||
XM_017012229.1 | 423 | UTR 3 | XP_016867718.1 | |||
XM_017012230.1 | 423 | UTR 3 | XP_016867719.1 |