Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTTCAGAACGAATTGACAAACA[A/C]ATTCGGTACATCCTCGACGGCATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147620 | ||||||||||||||||||||
Literature Links: |
IL6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL6 - interleukin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000600.4 | 622 | Missense Mutation | CAA,CAC | Q,H 56 | NP_000591.1 | |
NM_001318095.1 | 622 | Intron | NP_001305024.1 | |||
XM_005249745.4 | 622 | Missense Mutation | CAA,CAC | Q,H 110 | XP_005249802.1 | |
XM_011515390.2 | 622 | Missense Mutation | CAA,CAC | Q,H 56 | XP_011513692.1 |
LOC541472 - uncharacterized LOC541472 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |