Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTGCCTCTGGATGCGGCCTTT[A/G]CAGCGCTCGCGGTAGTCGGACTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186590 MIM: 615733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4284 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4284 - microRNA 4284 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX1A - syntaxin 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165903.1 | 506 | Silent Mutation | TGC,TGT | C,C 145 | NP_001159375.1 | |
NM_004603.3 | 506 | Silent Mutation | TGC,TGT | C,C 145 | NP_004594.1 | |
XM_017012567.1 | 506 | Silent Mutation | TGC,TGT | C,C 145 | XP_016868056.1 |
WBSCR22 - Williams-Beuren syndrome chromosome region 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |