Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCTGGAGATGCTCTGCATGGGT[A/G]CCGGGGGGGCTGGGGGAGGGCAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600197 | ||||||||||||||||||||
Literature Links: |
MAFK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAFK - MAF bZIP transcription factor K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM184A - transmembrane protein 184A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097620.1 | 2331 | Missense Mutation | GCA,GTA | A,V 341 | NP_001091089.1 | |
XM_006715653.1 | 2331 | Silent Mutation | GGC,GGT | G,G 360 | XP_006715716.1 | |
XM_006715655.2 | 2331 | Missense Mutation | GCA,GTA | A,V 320 | XP_006715718.1 | |
XM_011515164.1 | 2331 | Silent Mutation | GGC,GGT | G,G 340 | XP_011513466.1 | |
XM_011515165.2 | 2331 | Intron | XP_011513467.1 | |||
XM_011515166.2 | 2331 | Intron | XP_011513468.1 | |||
XM_017011815.1 | 2331 | UTR 5 | XP_016867304.1 |