Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGGCATAATTGCAAGTAATTT[C/T]GTTTTTCCTGCTACTCTTGTTCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606589 | ||||||||||||||||||||
Literature Links: |
SNX13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX13 - sorting nexin 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015132.4 | 2748 | Silent Mutation | NP_055947.1 | |||
XM_005249672.1 | 2748 | Silent Mutation | XP_005249729.1 | |||
XM_005249673.4 | 2748 | Intron | XP_005249730.1 | |||
XM_011515229.1 | 2748 | Silent Mutation | XP_011513531.1 | |||
XM_011515230.2 | 2748 | Silent Mutation | XP_011513532.1 | |||
XM_011515231.2 | 2748 | Silent Mutation | XP_011513533.1 | |||
XM_011515233.2 | 2748 | Silent Mutation | XP_011513535.1 | |||
XM_017011893.1 | 2748 | Silent Mutation | XP_016867382.1 | |||
XM_017011894.1 | 2748 | Silent Mutation | XP_016867383.1 | |||
XM_017011895.1 | 2748 | Silent Mutation | XP_016867384.1 | |||
XM_017011896.1 | 2748 | Silent Mutation | XP_016867385.1 | |||
XM_017011897.1 | 2748 | Silent Mutation | XP_016867386.1 |