Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGCTCACATGTCCCCGCCATCG[A/G]TGGGTCCCAGGACGCTGAGGCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF746 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF746 - zinc finger protein 746 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163474.1 | 2317 | Missense Mutation | ACC,ATC | T,I 640 | NP_001156946.1 | |
NM_152557.4 | 2317 | Missense Mutation | ACC,ATC | T,I 639 | NP_689770.3 | |
XM_005249955.2 | 2317 | Missense Mutation | ACC,ATC | T,I 655 | XP_005250012.1 | |
XM_005249956.2 | 2317 | Missense Mutation | ACC,ATC | T,I 654 | XP_005250013.1 | |
XM_005249957.1 | 2317 | Missense Mutation | ACC,ATC | T,I 642 | XP_005250014.1 | |
XM_011515838.1 | 2317 | Missense Mutation | ACC,ATC | T,I 627 | XP_011514140.1 |