Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGCCAAACAGCTGTTGTTGAGC[A/G]AATTGTTAGAACATCTTCTGGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600639 MIM: 616524 | ||||||||||||||||||||
Literature Links: |
CASP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASP2 - caspase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001224.4 | 158 | Missense Mutation | AAA,GAA | K,E 24 | NP_001215.1 | |
NM_032982.3 | 158 | Missense Mutation | AAA,GAA | K,E 55 | NP_116764.2 | |
NM_032983.3 | 158 | Missense Mutation | AAA,GAA | K,E 55 | NP_116765.2 |
LOC105375545 - uncharacterized LOC105375545 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM139 - transmembrane protein 139 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |