Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCTTTGCCTTCTCTACTTCTCA[C/G]TGATTGAACATAAGCTTCCAGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606821 MIM: 616714 | ||||||||||||||||||||
Literature Links: |
COG5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COG5 - component of oligomeric golgi complex 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161520.1 | 3022 | Intron | NP_001154992.1 | |||
NM_006348.3 | 3022 | Missense Mutation | CTG,GTG | L,V 833 | NP_006339.3 | |
NM_181733.2 | 3022 | Missense Mutation | CTG,GTG | L,V 812 | NP_859422.2 |
HBP1 - HMG-box transcription factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |