Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGAAATCAGCAAATACGTAAT[G/T]TACAAGTTCCTGACGGTGTTCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601048 | ||||||||||||||||||||
Literature Links: |
CAV2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAV2 - caveolin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206747.1 | 433 | Missense Mutation | ATG,ATT | M,I 71 | NP_001193676.1 | |
NM_001206748.1 | 433 | Missense Mutation | ATG,ATT | M,I 1 | NP_001193677.1 | |
NM_001233.4 | 433 | Missense Mutation | ATG,ATT | M,I 84 | NP_001224.1 | |
NM_198212.2 | 433 | Intron | NP_937855.1 |