Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAATGAAAAATACGGAGAAATGT[C/G]GGGGTGAGTATTCTGGAAACTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606534 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP3A43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP3A43 - cytochrome P450 family 3 subfamily A member 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278921.1 | 244 | Missense Mutation | CGG,GGG | R,G 113 | NP_001265850.1 | |
NM_022820.4 | 244 | Missense Mutation | TCG,TGG | S,W 72 | NP_073731.1 | |
NM_057095.2 | 244 | Missense Mutation | TCG,TGG | S,W 72 | NP_476436.1 | |
NM_057096.3 | 244 | Missense Mutation | TCG,TGG | S,W 72 | NP_476437.1 | |
XM_017012542.1 | 244 | UTR 5 | XP_016868031.1 | |||
XM_017012543.1 | 244 | UTR 5 | XP_016868032.1 | |||
XM_017012544.1 | 244 | Intron | XP_016868033.1 | |||
XM_017012545.1 | 244 | Intron | XP_016868034.1 |