Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGTGTGGGCTGTACACAAGAT[C/G]AGGGTCAAATAACCCAGTTTGGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611098 | ||||||||||||||||||||
Literature Links: |
STEAP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STEAP4 - STEAP4 metalloreductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205315.1 | 1396 | Silent Mutation | CTC,CTG | L,L 391 | NP_001192244.1 | |
NM_001205316.1 | 1396 | Silent Mutation | CTC,CTG | L,L 215 | NP_001192245.1 | |
NM_024636.3 | 1396 | Silent Mutation | CTC,CTG | L,L 391 | NP_078912.2 |