Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCATGAAAGCAGTGTGGAAACAG[A/G]ACAAAACCCCCAGAAAGGCAATCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605188 | ||||||||||||||||||||
Literature Links: |
GPR85 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR85 - G protein-coupled receptor 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146265.1 | 1000 | Missense Mutation | TCC,TTC | S,F 104 | NP_001139737.1 | |
NM_001146266.1 | 1000 | Missense Mutation | TCC,TTC | S,F 104 | NP_001139738.1 | |
NM_001146267.1 | 1000 | Missense Mutation | TCC,TTC | S,F 104 | NP_001139739.1 | |
NM_018970.6 | 1000 | Missense Mutation | TCC,TTC | S,F 104 | NP_061843.3 | |
XM_005250451.2 | 1000 | Missense Mutation | TCC,TTC | S,F 104 | XP_005250508.1 | |
XM_017012352.1 | 1000 | Missense Mutation | TCC,TTC | S,F 104 | XP_016867841.1 |