Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGATGCAGGCCCCCGCCACAAC[A/G]TCCACCCCACACAGGTAAAGTGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600639 MIM: 118425 | ||||||||||||||||||||
Literature Links: |
CASP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASP2 - caspase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLCN1 - chloride voltage-gated channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000083.2 | 253 | Missense Mutation | ATC,GTC | I,V 56 | NP_000074.2 | |
XM_011515782.2 | 253 | Intron | XP_011514084.1 | |||
XM_017011739.1 | 253 | Intron | XP_016867228.1 | |||
XM_017011740.1 | 253 | Intron | XP_016867229.1 |