Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTGTGACATATCAAAGTACTGA[G/T]AGAAGTTTTTCCTTGTCAGGTGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608910 | ||||||||||||||||||||
Literature Links: |
ARHGEF35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF35 - Rho guanine nucleotide exchange factor 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003702.2 | Intron | NP_001003702.2 |
CTAGE4 - CTAGE family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198495.2 | Intron | NP_940897.2 |
LOC101928605 - uncharacterized LOC101928605 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |