Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGATCCAGTTCATTGTTGGTGA[C/G]TTTCACCTTTTCAAAAGACACCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606061 | ||||||||||||||||||||
Literature Links: |
TBX20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBX20 - T-box 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077653.2 | 1102 | Missense Mutation | CTC,GTC | L,V 208 | NP_001071121.1 | |
NM_001166220.1 | 1102 | Missense Mutation | CTC,GTC | L,V 208 | NP_001159692.1 | |
XM_017012456.1 | 1102 | Missense Mutation | CTC,GTC | L,V 9 | XP_016867945.1 |