Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCTGGCTGCAGGAATACCGCCC[C/T]GATAGTGGAGAGGAGGCCGTGACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601260 | ||||||||||||||||||||
Literature Links: |
ZKSCAN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZKSCAN1 - zinc finger with KRAB and SCAN domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287054.1 | 606 | Silent Mutation | CCC,CCT | P,P 85 | NP_001273983.1 | |
NM_001287055.1 | 606 | Intron | NP_001273984.1 | |||
NM_003439.2 | 606 | Silent Mutation | CCC,CCT | P,P 121 | NP_003430.1 | |
XM_005250565.4 | 606 | Silent Mutation | CCC,CCT | P,P 121 | XP_005250622.1 | |
XM_011516559.2 | 606 | Silent Mutation | CCC,CCT | P,P 121 | XP_011514861.1 |