Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGATTTCAATGCGGCGGCGCCG[C/T]GTCAGGTAGCGGTTGAAGTGGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142954 MIM: 142951 MIM: 142950 MIM: 142956 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXA-AS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXA-AS3 - HOXA cluster antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA10-HOXA9 - HOXA10-HOXA9 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA3 - homeobox A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA6 - homeobox A6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA7 - homeobox A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006896.3 | 600 | Silent Mutation | NP_008827.2 |
HOXA9 - homeobox A9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |