Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGGGTACATTTTCTTCTGTGT[A/C]CTCAAAGCGTAGACTTTTGAATACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPG2 - coatomer protein complex subunit gamma 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSGA13 - testis specific 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304968.1 | 1443 | Missense Mutation | AGG,AGT | R,S 197 | NP_001291897.1 | |
NM_052933.3 | 1443 | Missense Mutation | AGG,AGT | R,S 197 | NP_443165.1 |