Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAACAAGCAGGACATGCCCAAC[A/G]CCATGCCCGTGAGCGAGCTGACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103188 MIM: 615800 MIM: 607418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARF5 - ADP ribosylation factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001662.3 | 544 | Missense Mutation | ACC,GCC | T,A 133 | NP_001653.1 |
FSCN3 - fascin actin-bundling protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCC1 - GRIP and coiled-coil domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |