Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTTGCTGCCTGTGATTCCTTCC[A/G]AGACATCACTCTCTCCGACTTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INMT - indolethylamine N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199219.1 | 249 | Missense Mutation | CAA,CGA | Q,R 78 | NP_001186148.1 | |
NM_006774.4 | 249 | Missense Mutation | CAA,CGA | Q,R 79 | NP_006765.4 |
INMT-FAM188B - INMT-FAM188B readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |