Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCACATTTTTAAATGGAAATAG[A/G]TAACCAGACGTGGGTGAGAGAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608497 | ||||||||||||||||||||
Literature Links: |
OR2F1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR2F1 - olfactory receptor family 2 subfamily F member 1 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012369.2 | 11 | Intron | NP_036501.2 | |||
XM_011516019.2 | 11 | Intron | XP_011514321.1 | |||
XM_011516020.2 | 11 | Intron | XP_011514322.1 |
OR2F2 - olfactory receptor family 2 subfamily F member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004685.1 | 11 | Missense Mutation | GAT,GGT | D,G 4 | NP_001004685.1 |