Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTGTTCGACCAGCTGGTTGAAG[C/T]GGGTTTCCGTCTTATTTCCCTTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612074 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGC27345 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGC27345 - uncharacterized protein MGC27345 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM28 - RNA binding motif protein 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166135.1 | 2076 | Missense Mutation | CAC,CGC | H,R 588 | NP_001159607.1 | |
NM_018077.2 | 2076 | Missense Mutation | CAC,CGC | H,R 729 | NP_060547.2 | |
XM_011516370.2 | 2076 | Missense Mutation | CAC,CGC | H,R 763 | XP_011514672.1 | |
XM_017012389.1 | 2076 | Missense Mutation | CAC,CGC | H,R 688 | XP_016867878.1 | |
XM_017012390.1 | 2076 | Missense Mutation | CAC,CGC | H,R 654 | XP_016867879.1 |