Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGACAATGTAATACCCTTGCCTGG[A/C]ATCCACTGGATAGTAACTGGCTAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615359 | ||||||||||||||||||||
Literature Links: |
LOC101927391 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927391 - uncharacterized LOC101927391 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIOS - meiosis regulator for oocyte development | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019005.3 | 498 | Missense Mutation | AAT,CAT | N,H 122 | NP_061878.3 | |
XM_005249780.3 | 498 | Missense Mutation | AAT,CAT | N,H 122 | XP_005249837.1 | |
XM_005249781.3 | 498 | Missense Mutation | AAT,CAT | N,H 122 | XP_005249838.1 | |
XM_005249782.3 | 498 | Missense Mutation | AAT,CAT | N,H 122 | XP_005249839.1 | |
XM_005249784.3 | 498 | Missense Mutation | AAT,CAT | N,H 122 | XP_005249841.1 | |
XM_011515432.2 | 498 | Missense Mutation | AAT,CAT | N,H 122 | XP_011513734.1 | |
XM_017012361.1 | 498 | Missense Mutation | AAT,CAT | N,H 122 | XP_016867850.1 | |
XM_017012362.1 | 498 | UTR 5 | XP_016867851.1 |