Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCAACCGCGGCTGTAGTGGAGG[G/T]GGAAGTAGAGCTTTTCCACCACGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC105375483 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105375483 - uncharacterized LOC105375483 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM229A - transmembrane protein 229A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136002.1 | 1356 | Missense Mutation | CAC,CCC | H,P 297 | NP_001129474.1 |