Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGGAGCCATGGCGGAGTCGGCG[C/T]CTGCTCGGGTAAAGAGGCACCGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602386 | ||||||||||||||||||||
Literature Links: |
ZNF212 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF212 - zinc finger protein 212 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012256.3 | 147 | Missense Mutation | CCT,TCT | P,S 6 | NP_036388.2 | |
XM_005250055.2 | 147 | Silent Mutation | CGC,CGT | R,R 47 | XP_005250112.2 | |
XM_011516581.2 | 147 | UTR 5 | XP_011514883.2 | |||
XM_011516582.1 | 147 | Intron | XP_011514884.1 | |||
XM_017012646.1 | 147 | Missense Mutation | CCT,TCT | P,S 6 | XP_016868135.1 | |
XM_017012647.1 | 147 | UTR 5 | XP_016868136.1 | |||
XM_017012648.1 | 147 | UTR 5 | XP_016868137.1 |