Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAGCGTGAAAGGGACAGAGACC[A/G]ACAACGAGAGGGGCATGGACGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614112 MIM: 187280 | ||||||||||||||||||||
Literature Links: |
LOC100507071 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100507071 - uncharacterized LOC100507071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR320A - microRNA 320a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3D - RNA polymerase III subunit D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001722.2 | 346 | Missense Mutation | CAA,CGA | Q,R 88 | NP_001713.2 |