Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGAATATGCGGCGCGCGTGGAT[A/C]CTGCTCACCTTGGGCTTGGTGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142460 | ||||||||||||||||||||
Literature Links: |
SDC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDC2 - syndecan 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002998.3 | 636 | Silent Mutation | ATA,ATC | I,I 6 | NP_002989.2 | |
XM_011517212.2 | 636 | Intron | XP_011515514.1 |