Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGACTGAATTTTCTGCTGTCCA[C/G]GAGCCATATCTAAAAACAGAAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF706 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF706 - zinc finger protein 706 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042510.1 | 308 | Missense Mutation | CCT,CGT | P,R 3 | NP_001035975.1 | |
NM_001267708.1 | 308 | Missense Mutation | CCT,CGT | P,R 3 | NP_001254637.1 | |
NM_001267709.1 | 308 | Missense Mutation | CCT,CGT | P,R 3 | NP_001254638.1 | |
NM_016096.3 | 308 | Missense Mutation | CCT,CGT | P,R 3 | NP_057180.1 |