Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACACGGCCTCCTGGGCCCGGCG[A/G]TACTCATAGACGCTGCCGTGCTCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614243 MIM: 610874 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6846 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6846 - microRNA 6846 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPLAH - 5-oxoprolinase (ATP-hydrolysing) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017570.4 | 3947 | Silent Mutation | TAC,TAT | Y,Y 1281 | NP_060040.1 | |
XM_011516960.1 | 3947 | Silent Mutation | TAC,TAT | Y,Y 1377 | XP_011515262.1 |
SMPD5 - sphingomyelin phosphodiesterase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017014128.1 | 3947 | Intron | XP_016869617.1 |
SPATC1 - spermatogenesis and centriole associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |