Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATTTGCTGGTTCCAGTTCCAAA[C/T]GGCATTAGGTAACACTTCAGCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603711 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP7B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP7B1 - cytochrome P450 family 7 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324112.1 | 1527 | Intron | NP_001311041.1 | |||
NM_004820.4 | 1527 | Silent Mutation | NP_004811.1 | |||
XM_017014002.1 | 1527 | Silent Mutation | XP_016869491.1 |