Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCCCGCATTTTAATGTTTTCAG[C/G]GGGGTGTCATAGCCCCGGGTTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603258 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKBKB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IKBKB - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190720.2 | 58 | Missense Mutation | GCG,GGG | A,G 4 | NP_001177649.1 | |
NM_001242778.1 | 58 | UTR 5 | NP_001229707.1 | |||
NM_001556.2 | 58 | UTR 5 | NP_001547.1 | |||
XM_005273490.2 | 58 | UTR 5 | XP_005273547.1 | |||
XM_005273491.4 | 58 | UTR 5 | XP_005273548.1 | |||
XM_005273492.3 | 58 | UTR 5 | XP_005273549.1 | |||
XM_005273493.3 | 58 | UTR 5 | XP_005273550.1 | |||
XM_005273494.2 | 58 | UTR 5 | XP_005273551.1 | |||
XM_005273495.2 | 58 | Intron | XP_005273552.1 | |||
XM_005273496.3 | 58 | UTR 5 | XP_005273553.1 | |||
XM_005273498.3 | 58 | UTR 5 | XP_005273555.1 | |||
XM_011544517.2 | 58 | Intron | XP_011542819.1 | |||
XM_011544518.2 | 58 | UTR 5 | XP_011542820.1 | |||
XM_011544519.2 | 58 | Intron | XP_011542821.1 | |||
XM_011544520.2 | 58 | Intron | XP_011542822.1 | |||
XM_011544521.2 | 58 | UTR 5 | XP_011542823.1 | |||
XM_011544522.2 | 58 | UTR 5 | XP_011542824.1 | |||
XM_017013396.1 | 58 | UTR 5 | XP_016868885.1 |
LOC101929897 - uncharacterized LOC101929897 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |