Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCTTCTCGCAGCAGTCCACGTC[C/G]ACCTTTAAGATCCCAGAGTTAATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603625 | ||||||||||||||||||||
Literature Links: |
LY6H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LY6H - lymphocyte antigen 6 complex, locus H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130478.1 | 466 | Silent Mutation | GTC,GTG | V,V 121 | NP_001123950.1 | |
NM_001135655.1 | 466 | Silent Mutation | GTC,GTG | V,V 121 | NP_001129127.1 | |
NM_002347.4 | 466 | Silent Mutation | GTC,GTG | V,V 100 | NP_002338.3 | |
XM_017013413.1 | 466 | Silent Mutation | GTC,GTG | V,V 126 | XP_016868902.1 | |
XM_017013414.1 | 466 | Silent Mutation | GTC,GTG | V,V 100 | XP_016868903.1 |