Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCCTGCAACATCCCAGCAGTG[A/G]ATTCCCATTTCATAATATGCATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602064 | ||||||||||||||||||||
Literature Links: |
IMPA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IMPA1 - inositol monophosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144878.1 | 976 | Silent Mutation | ATC,ATT | I,I 275 | NP_001138350.1 | |
NM_001144879.1 | 976 | Missense Mutation | TCC,TTC | S,F 180 | NP_001138351.1 | |
NM_005536.3 | 976 | Silent Mutation | ATC,ATT | I,I 216 | NP_005527.1 |