Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACTTGAGTAGCGTAATCACAAC[A/G]TGGTCATCTCGGCCTTGCCATAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609747 MIM: 605609 | ||||||||||||||||||||
Literature Links: |
ABRA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABRA - actin binding Rho activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139166.4 | 1176 | Silent Mutation | CAC,CAT | H,H 374 | NP_631905.1 |
OXR1 - oxidation resistance 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |