Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATAAGACTGTTGCAGATGTTCC[G/T]TTTCTCGATTTCTTGTGACCAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NIPAL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NIPAL2 - NIPA like domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321635.1 | 1251 | Missense Mutation | AAG,ACG | K,T 332 | NP_001308564.1 | |
NM_001321636.1 | 1251 | Intron | NP_001308565.1 | |||
NM_024759.2 | 1251 | Missense Mutation | AAG,ACG | K,T 332 | NP_079035.1 | |
XM_011517302.2 | 1251 | Intron | XP_011515604.1 |