Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCCTCGGGCTCATCGTATATGT[C/G]GTCAGGTCGAGGGGGCAGGGTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOK2 - docking protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317800.1 | 1046 | Missense Mutation | CAC,GAC | H,D 189 | NP_001304729.1 | |
NM_003974.3 | 1046 | Missense Mutation | CAC,GAC | H,D 343 | NP_003965.2 | |
NM_201349.2 | 1046 | Missense Mutation | CAC,GAC | H,D 189 | NP_958728.1 | |
XM_005273680.4 | 1046 | Missense Mutation | CAC,GAC | H,D 249 | XP_005273737.1 |