Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAAACAGATCTTGCGTATGCTG[A/G]AGAAGGACGATGTGGGTACCAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614112 MIM: 187280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507071 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100507071 - uncharacterized LOC100507071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR320A - microRNA 320a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3D - RNA polymerase III subunit D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001722.2 | 558 | Missense Mutation | AAG,GAG | K,E 159 | NP_001713.2 |