Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTGGAAACGGTTCAGAAATCT[A/G]TCCATTTTCCAAGAGCCTGGAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEBP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEBP4 - phosphatidylethanolamine binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144962.2 | 681 | Silent Mutation | GAC,GAT | D,D 178 | NP_659399.2 | |
XM_011544414.2 | 681 | Silent Mutation | GAC,GAT | D,D 178 | XP_011542716.1 | |
XM_017013103.1 | 681 | Intron | XP_016868592.1 |