Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCGTGTGGTTGGAGCTGGGCTC[C/T]TCCTCGGTGACCTGCGGGAGGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610931 | ||||||||||||||||||||
Literature Links: |
ZFAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFAT - zinc finger and AT-hook domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029939.3 | 3849 | Silent Mutation | GAA,GAG | E,E 1156 | NP_001025110.2 | |
NM_001167583.2 | 3849 | Silent Mutation | GAA,GAG | E,E 1156 | NP_001161055.1 | |
NM_001174157.1 | 3849 | Silent Mutation | GAA,GAG | E,E 1106 | NP_001167628.1 | |
NM_001174158.1 | 3849 | Silent Mutation | GAA,GAG | E,E 1070 | NP_001167629.1 | |
NM_001289394.1 | 3849 | Silent Mutation | GAA,GAG | E,E 1156 | NP_001276323.1 | |
NM_020863.3 | 3849 | Silent Mutation | GAA,GAG | E,E 1168 | NP_065914.2 | |
XM_011517203.1 | 3849 | Silent Mutation | GAA,GAG | E,E 1156 | XP_011515505.1 | |
XM_011517204.2 | 3849 | Silent Mutation | GAA,GAG | E,E 1106 | XP_011515506.1 | |
XM_011517206.1 | 3849 | Intron | XP_011515508.1 | |||
XM_017013716.1 | 3849 | Intron | XP_016869205.1 |