Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGGAATTAAGCATAAACTTAGA[A/T]ATATTGATCTACATAGTAATCGTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LRRCC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRCC1 - leucine rich repeat and coiled-coil centrosomal protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033402.4 | 284 | Missense Mutation | AAT,TAT | N,Y 139 | NP_208325.3 | |
XM_005251318.3 | 284 | Missense Mutation | AAT,TAT | N,Y 46 | XP_005251375.1 | |
XM_005251319.4 | 284 | UTR 5 | XP_005251376.1 | |||
XM_005251320.3 | 284 | UTR 5 | XP_005251377.1 | |||
XM_005251322.3 | 284 | UTR 5 | XP_005251379.1 | |||
XM_017013920.1 | 284 | Missense Mutation | AAT,TAT | N,Y 139 | XP_016869409.1 | |
XM_017013921.1 | 284 | Missense Mutation | AAT,TAT | N,Y 139 | XP_016869410.1 | |
XM_017013922.1 | 284 | Missense Mutation | AAT,TAT | N,Y 139 | XP_016869411.1 | |
XM_017013923.1 | 284 | Missense Mutation | AAT,TAT | N,Y 46 | XP_016869412.1 | |
XM_017013924.1 | 284 | Missense Mutation | AAT,TAT | N,Y 139 | XP_016869413.1 | |
XM_017013925.1 | 284 | UTR 5 | XP_016869414.1 | |||
XM_017013926.1 | 284 | UTR 5 | XP_016869415.1 | |||
XM_017013927.1 | 284 | UTR 5 | XP_016869416.1 | |||
XM_017013928.1 | 284 | UTR 5 | XP_016869417.1 |