Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCCTGACCAACTGCACGGAGCC[G/T]CTCAAGGCCATCGAGCAGTTCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf173-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf173-AS1 - C9orf173 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM166A - family with sequence similarity 166 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NELFB - negative elongation factor complex member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015456.4 | 288 | Silent Mutation | CCG,CCT | P,P 74 | NP_056271.3 |
STPG3 - sperm-tail PG-rich repeat containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |