Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAATGGCATTCATGTCAGGGAAA[C/T]TGAAGATTAAAGGTAACATGGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf147 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf147 - chromosome 9 open reading frame 147 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSDL2 - hydroxysteroid dehydrogenase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195822.1 | 1672 | Silent Mutation | CTG,TTG | L,L 322 | NP_001182751.1 | |
NM_032303.4 | 1672 | Silent Mutation | CTG,TTG | L,L 395 | NP_115679.2 | |
XM_011519091.2 | 1672 | Intron | XP_011517393.1 | |||
XM_017015203.1 | 1672 | Silent Mutation | CTG,TTG | L,L 221 | XP_016870692.1 | |
XM_017015204.1 | 1672 | Silent Mutation | CTG,TTG | L,L 221 | XP_016870693.1 | |
XM_017015205.1 | 1672 | Silent Mutation | CTG,TTG | L,L 221 | XP_016870694.1 |